ContactUsLinkComponent
Human IL-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human IL-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA3578-96
Supplier:  Antibodies.com
New Product
Human IL-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human IL-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA3578-96
Supplier:  Antibodies.com
New Product