ContactUsLinkComponent
Human Leukotriene A4 Hydrolase/LTA4H ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Leukotriene A4 Hydrolase/LTA4H in serum, plasma, tissue homogenates or other biolog 1 * 96 Tests
Catalog # ANTIA5302-96
Supplier:  ANTIBODIES.COM
New Product
Human Leukotriene A4 Hydrolase/LTA4H ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Leukotriene A4 Hydrolase/LTA4H in serum, plasma, tissue homogenates or other biolog 1 * 96 Tests
Catalog # ANTIA5302-96
Supplier:  ANTIBODIES.COM
New Product