Mouse CD167a/DDR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse CD167a/DDR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA4847-96
Supplier: Antibodies.com
New Product