ContactUsLinkComponent
Human Extracellular Matrix Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Extracellular Matrix Protein 1 in serum, plasma, tissue homogenates, cell lysates, 1 * 48 Tests
Catalog # ANTIA4224-48
Supplier:  ANTIBODIES.COM
New Product
Human Extracellular Matrix Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Extracellular Matrix Protein 1 in serum, plasma, tissue homogenates, cell lysates, 1 * 48 Tests
Catalog # ANTIA4224-48
Supplier:  ANTIBODIES.COM
New Product