ContactUsLinkComponent
Mouse GABA A Receptor alpha 2/GABRA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse GABA A Receptor alpha 2/GABRA2 in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA5514-48
Supplier:  ANTIBODIES.COM
New Product
Mouse GABA A Receptor alpha 2/GABRA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse GABA A Receptor alpha 2/GABRA2 in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA5514-48
Supplier:  ANTIBODIES.COM
New Product