ContactUsLinkComponent
Human ANGPTL2/ARP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human ANGPTL2/ARP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA3687-96
Supplier:  Antibodies.com
New Product
Human ANGPTL2/ARP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human ANGPTL2/ARP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA3687-96
Supplier:  Antibodies.com
New Product