Human Von Hippel Lindau/VHL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Von Hippel Lindau/VHL in serum, plasma, tissue homogenates, cell lysates, cell culture super 1 * 96 Tests
Catalog # ANTIA82324-96
Supplier: Antibodies.com
New Product