ContactUsLinkComponent
Monkey Thrombin/Antithrombin Complex ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of monkey Thrombin/Antithrombin Complex in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA76019-96
Supplier:  ANTIBODIES.COM
New Product
Monkey Thrombin/Antithrombin Complex ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of monkey Thrombin/Antithrombin Complex in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA76019-96
Supplier:  ANTIBODIES.COM
New Product