ContactUsLinkComponent
Rat HSP90 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat HSP90 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA6931-96
Supplier:  Antibodies.com
New Product
Rat HSP90 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat HSP90 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA6931-96
Supplier:  Antibodies.com
New Product