ContactUsLinkComponent
Human Carbonic Anhydrase 9/CA9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 9/CA9 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7162-96
Supplier:  Antibodies.com
New Product
Human Carbonic Anhydrase 9/CA9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 9/CA9 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7162-96
Supplier:  Antibodies.com
New Product