ContactUsLinkComponent
Rat Xanthine Oxidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Xanthine Oxidase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7546-96
Supplier:  Antibodies.com
New Product
Rat Xanthine Oxidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Xanthine Oxidase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7546-96
Supplier:  Antibodies.com
New Product