ContactUsLinkComponent
Mouse Leukotriene B4 Receptor 1/BLT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Leukotriene B4 Receptor 1/BLT in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA76907-96
Supplier:  Antibodies.com
New Product
Mouse Leukotriene B4 Receptor 1/BLT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Leukotriene B4 Receptor 1/BLT in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA76907-96
Supplier:  Antibodies.com
New Product