ContactUsLinkComponent
Monkey Carbonic Anhydrase 2/CA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of monkey Carbonic Anhydrase 2/CA2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6964-48
Supplier:  Antibodies.com
New Product
Monkey Carbonic Anhydrase 2/CA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of monkey Carbonic Anhydrase 2/CA2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6964-48
Supplier:  Antibodies.com
New Product