ContactUsLinkComponent
Rat NUCB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat NUCB2 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA7515-96
New Product
Rat NUCB2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat NUCB2 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA7515-96
New Product