ContactUsLinkComponent
Mouse Trypsinogen Activation Peptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Trypsinogen Activation Peptide in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79733-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Trypsinogen Activation Peptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Trypsinogen Activation Peptide in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79733-96
Supplier:  ANTIBODIES.COM
New Product