ContactUsLinkComponent
Rat GRO gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat GRO gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA7166-48
Supplier:  Antibodies.com
New Product
Rat GRO gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat GRO gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA7166-48
Supplier:  Antibodies.com
New Product