ContactUsLinkComponent
Human Extracellular Matrix Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Extracellular Matrix Protein 1 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78022-96
Supplier:  ANTIBODIES.COM
New Product
Human Extracellular Matrix Protein 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Extracellular Matrix Protein 1 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78022-96
Supplier:  ANTIBODIES.COM
New Product