ContactUsLinkComponent
Rat DIO2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat DIO2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6509-96
Supplier:  Antibodies.com
New Product
Rat DIO2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat DIO2 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6509-96
Supplier:  Antibodies.com
New Product