ContactUsLinkComponent
Mouse Smad3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Smad3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7074-48
Supplier:  Antibodies.com
New Product
Mouse Smad3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Smad3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7074-48
Supplier:  Antibodies.com
New Product