ContactUsLinkComponent
Chicken alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of chicken alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA87097-96
Supplier:  Antibodies.com
New Product
Chicken alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of chicken alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA87097-96
Supplier:  Antibodies.com
New Product