ContactUsLinkComponent
Mouse Relaxin 3 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of mouse Relaxin 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
Catalog # ANTIA82368-96
Supplier:  Antibodies.com
New Product
Mouse Relaxin 3 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of mouse Relaxin 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
Catalog # ANTIA82368-96
Supplier:  Antibodies.com
New Product