ContactUsLinkComponent
Human Ornithine Aminotransferase/OAT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Ornithine Aminotransferase/OAT in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79581-96
Supplier:  ANTIBODIES.COM
New Product
Human Ornithine Aminotransferase/OAT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Ornithine Aminotransferase/OAT in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79581-96
Supplier:  ANTIBODIES.COM
New Product