ContactUsLinkComponent
Mouse GIP/Gastric Inhibitory Peptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse GIP/Gastric Inhibitory Peptide in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79381-96
Supplier:  ANTIBODIES.COM
New Product
Mouse GIP/Gastric Inhibitory Peptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse GIP/Gastric Inhibitory Peptide in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA79381-96
Supplier:  ANTIBODIES.COM
New Product