Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tin(II) chloride dihydrate ACS reagent, 98% 1 * 100 g
Catalog # SIAL243523-100G
Supplier:
Merck
New Product
Expand image
Tin(II) chloride dihydrate ACS reagent, 98% 1 * 100 g
Catalog # SIAL243523-100G
Supplier:
Merck
New Product