0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,3-Diethoxy-1-propanol
BP0000702159
3,3-Diethoxypropan-1-ol
Bottle
APOSOR74677-100G
New Product
:
Apollo Scientific
3,3-Diethoxypropan-1-ol
APOSOR74677-100G
:
Apollo Scientific
New Product
:
3,3-Diethoxypropan-1-ol
Bottle
:
1 100,00 €
1 * 100 g
-
+
Pack type:
Bottle