0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Propanedithiol
1,3-Propanedithiol 97%
1,3-Propanedithiol 97%
1,3-Propanedithiol 97%
Bottle
APOSOR70117-500G
New Product
:
Apollo Scientific
1,3-Propanedithiol 97%
APOSOR70117-500G
:
Apollo Scientific
New Product
:
109-80-8
Expand image
1,3-Propanedithiol 97%
Bottle
Add to cart
:
109-80-8
346,00 €
Unit of Measure
1 * 500 g
-
+
Add to cart
:
1,3-Dimercaptopropane
:
109-80-8
:
C₃H₈S₂
:
MFCD00004904
:
108.23 g/mol
:
168…170 °C
:
–79 °C
:
1.077 g/cm³ (25 °C)
:
63 °C
:
Refrigerator
:
3334
:
9,III
Pack type:
Bottle