Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chlorophenylurea 98%

4-Chlorophenylurea 98%

Bottle
Catalog # APOSOR61200-100G
Supplier:   Apollo Scientific
4-Chlorophenylurea 98%
Catalog # APOSOR61200-100G
Supplier:  Apollo Scientific
CAS Number:  140-38-5
4-Chlorophenylurea 98%

Bottle

CAS Number:  140-38-5
Price
400,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-(4-Chlorophenyl)urea
  • CAS Number:140-38-5
  • Formula:C₇H₇ClN₂O
  • MDL Number:MFCD00014788
  • Molecular Weight:170.6 g/mol
  • Melting Pt:205…205 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications