0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,4-Dimethylbenzophenone
BP0000711479
3,4-Dimethylbenzophenone
Bottle
APOSOR72539-500G
New Product
:
Apollo Scientific
3,4-Dimethylbenzophenone
APOSOR72539-500G
:
Apollo Scientific
New Product
:
3,4-Dimethylbenzophenone
Bottle
:
112,00 €
1 * 500 g
-
+
Pack type:
Bottle