Order Entry
Export
Orders LinkContactUsLinkComponent
Triphenylsilane 96%

Triphenylsilane 96%

Bottle
Catalog # APOSOR72474-100G
New Product
Supplier:   Apollo Scientific
Triphenylsilane 96%
Catalog # APOSOR72474-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  789-25-3
Triphenylsilane 96%

Bottle

CAS Number:  789-25-3
Price
116,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • Store at 2 to 8 °C
  • CAS Number:789-25-3
  • Formula:C₁₈H₁₆Si
  • MDL Number:MFCD00003003
  • Molecular Weight:260,41 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:212-333-0
Specifications

Specifications

  • Pack type:
    Bottle

Specifications