Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethylphenylsilane 97%

Dimethylphenylsilane 97%

Bottle
Catalog # APOSOR72472-100G
Flammable
Irritant
Supplier:   Apollo Scientific
Dimethylphenylsilane 97%
Catalog # APOSOR72472-100G
Supplier:  Apollo Scientific
CAS Number:  766-77-8
Dimethylphenylsilane 97%

Bottle

CAS Number:  766-77-8
Price
116,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Dimethyl(phenyl)silane
  • CAS Number:766-77-8
  • Formula:C₈H₁₂Si
  • MDL Number:MFCD00008257
  • Molecular Weight:136,27 g/mol
  • EINECS:212-170-5
  • UN:1993
Specifications

Specifications

  • Pack type:
    Bottle

Specifications