Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Diphenylsilane
BP0000755328
Print
Diphenylsilane
Bottle
Catalog # APOSOR72471-100G
New Product
Supplier:
Apollo Scientific
Diphenylsilane
Catalog # APOSOR72471-100G
Supplier:
Apollo Scientific
New Product
CAS Number:
Return to Top
Diphenylsilane
Bottle
Specifications
Jump to:
Specifications
CAS Number:
Price
176,00 €
Log in
for availability
1 * 100 g
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications