4,7-Dihydroxyquinazoline
Bottle
Catalog # APOSOR471457-250MG
New Product
Supplier: Apollo Scientific
4,7-Dihydroxyquinazoline
Catalog # APOSOR471457-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 16064-25-8
Jump to:
Product Details & Documents
- CAS Number:16064-25-8
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD01823180
- Molecular Weight:162.145 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



