Pentane-1,5-dithiol 98%
Bottle
Catalog # APOSOR42200-500G
New Product
Supplier: Apollo Scientific
Pentane-1,5-dithiol 98%
Catalog # APOSOR42200-500G
Supplier: Apollo Scientific
New Product
CAS Number: 928-98-3
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 1,5-Dimercaptopentane
- CAS Number:928-98-3
- Formula:C₅H₁₂S₂
- MDL Number:MFCD00004908
- Molecular Weight:136,27877 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR42200-500G



