Order Entry
Export
Orders LinkContactUsLinkComponent
3-Acetylbenzophenone

3-Acetylbenzophenone

Bottle
Catalog # APOSOR40003-25G
Irritant
Supplier:   Apollo Scientific
3-Acetylbenzophenone
Catalog # APOSOR40003-25G
Supplier:  Apollo Scientific
CAS Number:  66067-44-5
3-Acetylbenzophenone

Bottle

CAS Number:  66067-44-5
Price
8 660,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone
  • CAS Number:66067-44-5
  • Formula:C₁₅H₁₂O₂
  • MDL Number:MFCD01662280
  • Molecular Weight:224.2546 g/mol
  • Boiling Pt:80…85 °C/0.1mmHg
  • Density:1.22 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications