Order Entry
Export
Orders LinkContactUsLinkComponent
Tetracosane 99%

Tetracosane 99%

Bottle
Catalog # APOSOR1015180-100G
Supplier:   Apollo Scientific
Tetracosane 99%
Catalog # APOSOR1015180-100G
Supplier:  Apollo Scientific
CAS Number:  646-31-1
Tetracosane 99%

Bottle

CAS Number:  646-31-1
Price
228,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

  • Non-hazardous
Synonyms:  N-tetracosane
  • CAS Number:646-31-1
  • Formula:C₂₄H₅₀
  • MDL Number:MFCD00009352
  • Molecular Weight:338,65 g/mol
  • EINECS:211-474-5
Specifications

Specifications

  • Pack type:
    Bottle

Specifications