Order Entry
Export
Orders LinkContactUsLinkComponent
Perylene 98%

Perylene 98%

Bottle
Catalog # APOSOR1014604-100G
Irritant
Supplier:   Apollo Scientific
Perylene 98%
Catalog # APOSOR1014604-100G
Supplier:  Apollo Scientific
CAS Number:  198-55-0
Perylene 98%

Bottle

CAS Number:  198-55-0
Price
199,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:198-55-0
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00004142
  • Molecular Weight:252,31 g/mol
  • EINECS:205-900-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSOR1014604-100G

Specifications