Order Entry
Export
Orders LinkContactUsLinkComponent
 

N-Tetradecanamide 97%

Bottle
Catalog # APOSOR1014492-100G
Supplier:   Apollo Scientific
N-Tetradecanamide 97%
Catalog # APOSOR1014492-100G
Supplier:  Apollo Scientific
CAS Number:  638-58-4
N-Tetradecanamide 97%

Bottle

CAS Number:  638-58-4
Price
582,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  N-Tetradecanamide
  • CAS Number:638-58-4
  • Formula:C₁₄H₂₉NO
  • MDL Number:MFCD00025533
  • Molecular Weight:227,39 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications