Order Entry
Export
Orders LinkContactUsLinkComponent
Nonadecan-10-o 98%

Nonadecan-10-o 98%

Bottle
Catalog # APOSOR1014447-1G
New Product
Supplier:   Apollo Scientific
Nonadecan-10-o 98%
Catalog # APOSOR1014447-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  16840-84-9
Nonadecan-10-o 98%

Bottle

CAS Number:  16840-84-9
Price
141,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Nonadecan-10-ol, 10-Nonadecyl alcohol, 10-Hydroxy nonadecane
  • CAS Number:16840-84-9
  • Formula:C₁₉H₄₀O
  • MDL Number:MFCD00968670
  • Molecular Weight:284,52 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications