Order Entry
Export
Orders LinkContactUsLinkComponent
1,1-Diphenylethylene 98%

1,1-Diphenylethylene 98%

Bottle
Catalog # APOSOR315713-100G
New Product
Supplier:   Apollo Scientific
1,1-Diphenylethylene 98%
Catalog # APOSOR315713-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  
1,1-Diphenylethylene 98%

Bottle

CAS Number:  
Price
121,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
  • Molecular Weight:180,24508 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications