Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorobenzhydrazide

2,4-Dichlorobenzhydrazide

Bottle
Catalog # APOSOR311059-25G
Irritant
New Product
Supplier:   Apollo Scientific
2,4-Dichlorobenzhydrazide
Catalog # APOSOR311059-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  5814-06-2
2,4-Dichlorobenzhydrazide

Bottle

CAS Number:  5814-06-2
Price
808,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,4-Dichlorobenzoic hydrazide
  • CAS Number:5814-06-2
  • Formula:C₇H₆Cl₂N₂O
  • MDL Number:MFCD00025113
  • Molecular Weight:205.04 g/mol
  • Melting Pt:168…170 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications