Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,4-Dinitrobenzaldehyde
3,4-Dinitrobenzaldehyde
3,4-Dinitrobenzaldehyde
Bottle
APOSOR303581-5G
New Product
:
Apollo Scientific
3,4-Dinitrobenzaldehyde
APOSOR303581-5G
:
Apollo Scientific
New Product
:
35998-98-2
3,4-Dinitrobenzaldehyde
Bottle
:
35998-98-2
1 510,00 €
1 * 5 g
-
+
:
35998-98-2
:
C₇H₄N₂O₅
:
MFCD00845054
:
196.117 g/mol
:
83…85 °C
:
Ambient
Pack type:
Bottle