Order Entry
Export
Orders LinkContactUsLinkComponent
3-Hydroxyazetidine 98%

3-Hydroxyazetidine 98%

Bottle
Catalog # APOSOR301160-5G
Irritant
Supplier:   Apollo Scientific
3-Hydroxyazetidine 98%
Catalog # APOSOR301160-5G
Supplier:  Apollo Scientific
CAS Number:  45347-82-8
3-Hydroxyazetidine 98%

Bottle

CAS Number:  45347-82-8
Price
490,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3-Azetidinol, Azetidin-3-ol
  • CAS Number:45347-82-8
  • Formula:C₃H₇NO
  • MDL Number:MFCD03695446
  • Molecular Weight:73,09377 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications