3-Hydroxyazetidine 98%
Bottle
Catalog # APOSOR301160-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 3-Azetidinol, Azetidin-3-ol
- CAS Number:45347-82-8
- Formula:C₃H₇NO
- MDL Number:MFCD03695446
- Molecular Weight:73,09377 g/mol
Specifications
Specifications
- Pack type:Bottle



