Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
6-Methoxyoxindole
6-Methoxyoxindole
6-Methoxyoxindole
Bottle
APOSOR301018-250MG
:
Apollo Scientific
6-Methoxyoxindole
APOSOR301018-250MG
:
Apollo Scientific
:
7699-19-6
6-Methoxyoxindole
Bottle
:
7699-19-6
138,00 €
1 * 250 mg
-
+
:
7699-19-6
:
C₉H₉NO₂
:
MFCD00239414
:
163.18 g/mol
:
160…161 °C
:
Refrigerator
Pack type:
Bottle