Order Entry
Export
Orders LinkContactUsLinkComponent
 

6-Methoxyoxindole

Bottle
Catalog # APOSOR301018-10G
Irritant
Supplier:   Apollo Scientific
6-Methoxyoxindole
Catalog # APOSOR301018-10G
Supplier:  Apollo Scientific
CAS Number:  7699-19-6
6-Methoxyoxindole

Bottle

CAS Number:  7699-19-6
Price
1 590,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:7699-19-6
  • Formula:C₉H₉NO₂
  • MDL Number:MFCD00239414
  • Molecular Weight:163.18 g/mol
  • Melting Pt:160…161 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications