Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,4-Dimethoxybenzonitrile
3,4-Dimethoxybenzonitrile
3,4-Dimethoxybenzonitrile
3,4-Dimethoxybenzonitrile
Bottle
APOSOR2992-500G
New Product
:
Apollo Scientific
3,4-Dimethoxybenzonitrile
APOSOR2992-500G
:
Apollo Scientific
New Product
:
2024-83-1
Expand image
3,4-Dimethoxybenzonitrile
Bottle
Add to cart
:
2024-83-1
409,00 €
Unit of Measure
1 * 500 g
-
+
Add to cart
:
Veratronitrile
:
2024-83-1
:
C₉H₉NO₂
:
MFCD00001802
:
163.18 g/mol
:
65…70 °C
:
Ambient
:
217-969-2
:
3439
:
6.1,III
Pack type:
Bottle