Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxyquinazoline 98%

4-Hydroxyquinazoline 98%

Bottle
Catalog # APOSOR28898-500G
Irritant
Supplier:   Apollo Scientific
4-Hydroxyquinazoline 98%
Catalog # APOSOR28898-500G
Supplier:  Apollo Scientific
CAS Number:  491-36-1
4-Hydroxyquinazoline 98%

Bottle

CAS Number:  491-36-1
Price
157,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  4-Hydroxyquinazoline, Quinazolin-4(3H)-one
  • CAS Number:491-36-1
  • Formula:C₈H₆N₂O
  • MDL Number:MFCD00511302
  • Molecular Weight:146,14604 g/mol
  • EINECS:207-735-8
Specifications

Specifications

  • Pack type:
    Bottle

Specifications