Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl 5,6-dichloronicotinate 97%

Methyl 5,6-dichloronicotinate 97%

Bottle
Catalog # APOSOR27409-100G
Irritant
New Product
Supplier:   Apollo Scientific
Methyl 5,6-dichloronicotinate 97%
Catalog # APOSOR27409-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  56055-54-0
Methyl 5,6-dichloronicotinate 97%

Bottle

CAS Number:  56055-54-0
Price
453,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methyl 5,6-dichloronicotinate
  • CAS Number:56055-54-0
  • Formula:C₇H₅Cl₂NO₂
  • MDL Number:MFCD00125367
  • Molecular Weight:206.03 g/mol
  • Melting Pt:68…70 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications