Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,5-Dibromonaphthalene
1,5-Dibromonaphthalene 98%
1,5-Dibromonaphthalene 98%
Bottle
APOSOR16492-25G
New Product
:
Apollo Scientific
1,5-Dibromonaphthalene 98%
APOSOR16492-25G
:
Apollo Scientific
New Product
:
7351-74-8
1,5-Dibromonaphthalene 98%
Bottle
:
7351-74-8
623,00 €
1 * 25 g
-
+
:
7351-74-8
:
C₁₀H₆Br₂
:
MFCD00089984
:
285.9600 g/mol
:
130…130.5 °C
:
Ambient
Pack type:
Bottle