Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxycoumarin 98%

7-Hydroxycoumarin 98%

Bottle
Catalog # APOSOR14329-25G
Irritant
Supplier:   Apollo Scientific
7-Hydroxycoumarin 98%
Catalog # APOSOR14329-25G
Supplier:  Apollo Scientific
CAS Number:  93-35-6
7-Hydroxycoumarin 98%

Bottle

CAS Number:  93-35-6
Price
42,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162,14213 g/mol
  • EINECS:202-240-3
Specifications

Specifications

  • Pack type:
    Bottle

Specifications